3-(2-Chloro-5-methylpyridin-3-yl)prop-2-yn-1-ol

Supplier: Matrix Scientific

054081-100MG 054081-500MG
101906-090EA 364.62 USD
101906-090 101906-092
3-(2-Chloro-5-methylpyridin-3-yl)prop-2-yn-1-ol
3-(2-Chloro-5-methylpyridin-3-yl)prop-2-yn-1-ol

Hazard: Irritant

Formula: C₉H₈ClNO
MDL Number: MFCD13563048
CAS Number: 1203499-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR