2-Chloro-6-iodo-5-methylpyridin-3-ol
Supplier: Matrix Scientific
054080-100MG
054080-500MG
101906-086EA
280
USD
101906-086
101906-088
2-Chloro-6-iodo-5-methylpyridin-3-ol
2-Chloro-6-iodo-5-methylpyridin-3-ol
Hazard: Irritant
Formula:
C₆H₅ClINO |
MDL Number:
MFCD13563047 CAS Number: 1203499-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...