2-Chloro-3-methoxy-5-methylpyridine

Supplier: Matrix Scientific

054079-100MG 054079-500MG
101906-082EA 315.38 USD
101906-082 101906-084
2-Chloro-3-methoxy-5-methylpyridine
2-Chloro-3-methoxy-5-methylpyridine

Hazard: Irritant

Formula: C₇H₈ClNO
MDL Number: MFCD13563046
CAS Number: 1203499-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR