2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-methylpyridine-3-boronic acid pinacol ester

054075-100MG 054075-500MG
101906-066EA 364.62 USD
101906-066 101906-068
2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇BClNO₂
MDL Number: MFCD08063083
CAS Number: 1309982-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR