2-Chloro-5-methylpyridin-3-ol

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-hydroxy-5-methylpyridine, 2-Chloro-3-hydroxy-5-picoline

054074-100MG 054074-500MG
101906-062EA 269.23 USD
101906-062 101906-064
2-Chloro-5-methylpyridin-3-ol
2-Chloro-5-methylpyridin-3-ol

Hazard: Irritant

Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD09839272
CAS Number: 910649-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR