2-Chloro-5-methylnicotinaldehyde

Supplier: Matrix Scientific

054073-100MG 054073-500MG
101906-058EA 364.62 USD
101906-058 101906-060
2-Chloro-5-methylnicotinaldehyde
2-Chloro-5-methylnicotinaldehyde

Hazard: Irritant

Formula: C₇H₆ClNO
MDL Number: MFCD07375370
CAS Number: 92444-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR