N-(5-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)pivalamide

Supplier: Matrix Scientific

054068-100MG 054068-500MG
101906-038EA 364.62 USD
101906-038 101906-040
N-(5-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)pivalamide
N-(5-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₇H₂₇BN₂O₃
MDL Number: MFCD13563040
CAS Number: 1310383-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR