2-Amino-5-methylnicotinaldehyde

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-methylpyridine-3-carboxaldehyde

054066-100MG 054066-500MG
101906-030EA 92.31 USD
101906-030 101906-032
2-Amino-5-methylnicotinaldehyde
2-Amino-5-methylnicotinaldehyde

Hazard: Irritant

Formula: C₇H₈N₂O
MDL Number: MFCD09754122
CAS Number: 1023814-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR