N-(3-Formyl-5-methylpyridin-2-yl)pivalamide

Supplier: Matrix Scientific

054065-100MG 054065-500MG
101906-026EA 364.62 USD
101906-026 101906-028
N-(3-Formyl-5-methylpyridin-2-yl)pivalamide
N-(3-Formyl-5-methylpyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD13176641
CAS Number: 127446-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR