3-(Benzyloxy)-2-(2-(benzyloxy)ethoxy)pyridine, Technical Grade

Supplier: Matrix Scientific

054063-100MG 054063-500MG
101906-018EA 364.62 USD
101906-018 101906-020
3-(Benzyloxy)-2-(2-(benzyloxy)ethoxy)pyridine, Technical Grade
3-(Benzyloxy)-2-(2-(benzyloxy)ethoxy)pyridine

Hazard: Irritant

Formula: C₂₁H₂₁NO₃
MDL Number: MFCD13176640
CAS Number: 156840-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR