4-Chloro-N-phenylpicolinamide

Supplier: Matrix Scientific

054061-100MG 054061-500MG
101906-010EA 364.62 USD
101906-010 101906-012
4-Chloro-N-phenylpicolinamide
4-Chloro-N-phenylpicolinamide

Hazard: Irritant

Formula: C₁₂H₉ClN₂O
MDL Number: MFCD08523032
CAS Number: 133928-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR