3-Iodo-5-methylpyridin-2-ol

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-3-iodo-5-methylpyridine

054059-100MG 054059-500MG
101906-002EA 364.62 USD
101906-002 101906-004
3-Iodo-5-methylpyridin-2-ol
3-Iodo-5-methylpyridin-2-ol

Hazard: Irritant

Formula: C₆H₆INO
MW: 235.03 g/mol
MDL Number: MFCD09037469
CAS Number: 313678-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR