3-Bromo-5-nitropyridin-4-ol

Supplier: Matrix Scientific

054055-1G 054055-5G
101905-986EA 83.08 USD
101905-986 101905-988
3-Bromo-5-nitropyridin-4-ol
3-Bromo-5-nitropyridin-4-ol

Hazard: Irritant

Formula: C₅H₃BrN₂O₃
MDL Number: MFCD08692339
CAS Number: 31872-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR