3-(4-Chloropyridin-3-yl)prop-2-yn-1-ol

Supplier: Matrix Scientific

054054-100MG 054054-500MG
101905-982EA 761.54 USD
101905-982 101905-984
3-(4-Chloropyridin-3-yl)prop-2-yn-1-ol
3-(4-Chloropyridin-3-yl)prop-2-yn-1-ol

Hazard: Irritant

Formula: C₈H₆ClNO
MDL Number: MFCD13176636
CAS Number: 1203499-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR