N-(4-Hydroxy-5-((trimethylsilyl)ethynyl)pyridin-3-yl)acetamide

Supplier: Matrix Scientific

054053-100MG 054053-500MG
101905-978EA 364.62 USD
101905-978 101905-980
N-(4-Hydroxy-5-((trimethylsilyl)ethynyl)pyridin-3-yl)acetamide
N-(4-Hydroxy-5-((trimethylsilyl)ethynyl)pyridin-3-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂Si
MW: 248.36 g/mol
MDL Number: MFCD13176635
CAS Number: 1203499-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR