tert-Butyl-3-((5-bromo-3-formylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Supplier: Matrix Scientific

054052-100MG 054052-500MG
101905-974EA 364.62 USD
101905-974 101905-976
tert-Butyl-3-((5-bromo-3-formylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate
tert-Butyl-3-((5-bromo-3-formylpyridin-2-yloxy)methyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₁BrN₂O₄
MW: 385.27 g/mol
MDL Number: MFCD13176634
CAS Number: 1203499-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR