N-(2-Chloro-6-((3-(hydroxymethyl)pyrrolidin-1-yl)methyl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

054049-100MG 054049-500MG
101905-962EA 364.62 USD
101905-962 101905-964
N-(2-Chloro-6-((3-(hydroxymethyl)pyrrolidin-1-yl)methyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-((3-(hydroxymethyl)pyrrolidin-1-yl)methyl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₆H₂₄ClN₃O₂
MW: 325.84 g/mol
MDL Number: MFCD13176632
CAS Number: 1203499-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR