6-Bromo-2-chloro-4-((trimethylsilyl)ethynyl)pyridin-3-amine

Supplier: Matrix Scientific

054047-100MG 054047-500MG
101905-954EA 364.62 USD
101905-954 101905-956
6-Bromo-2-chloro-4-((trimethylsilyl)ethynyl)pyridin-3-amine
6-Bromo-2-chloro-4-((trimethylsilyl)ethynyl)pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₂BrClN₂Si
MW: 303.67 g/mol
MDL Number: MFCD13176630
CAS Number: 1203499-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR