N,N-Diisopropyl-4-methoxypicolinamide

Supplier: Matrix Scientific

054046-100MG 054046-500MG
101905-950EA 364.62 USD
101905-950 101905-952
N,N-Diisopropyl-4-methoxypicolinamide
N,N-Diisopropyl-4-methoxypicolinamide

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD13176629
CAS Number: 446019-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR