4-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine

Supplier: Matrix Scientific

054045-100MG 054045-500MG
101905-946EA 364.62 USD
101905-946 101905-948
4-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine
4-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-3-iodopyridine

Hazard: Irritant

Formula: C₁₆H₂₇IN₂OSi
MDL Number: MFCD13176628
CAS Number: 1203498-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR