N-(2-Bromo-5-iodopyridin-3-yl)pivalamide

Supplier: Matrix Scientific

054043-100MG 054043-500MG
101905-938EA 364.62 USD
101905-938 101905-940
N-(2-Bromo-5-iodopyridin-3-yl)pivalamide
N-(2-Bromo-5-iodopyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₂BrIN₂O
MDL Number: MFCD13176626
CAS Number: 1203498-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR