tert-Butyl-3,4-dichloropyridin-2-ylcarbamate

Supplier: Matrix Scientific

054041-100MG 054041-500MG
101905-930EA 364.62 USD
101905-930 101905-932
tert-Butyl-3,4-dichloropyridin-2-ylcarbamate
tert-Butyl-3,4-dichloropyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂N₂O₂
MW: 263.13 g/mol
MDL Number: MFCD13563037
CAS Number: 1203499-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR