N-(2-Chloro-3-methylpyridin-4-yl)pivalamide

Supplier: Matrix Scientific

054040-100MG 054040-500MG
101905-926EA 364.62 USD
101905-926 101905-928
N-(2-Chloro-3-methylpyridin-4-yl)pivalamide
N-(2-Chloro-3-methylpyridin-4-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD13176624
CAS Number: 1203499-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR