3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5,6-dihydropyridine-1(2H)-carboxylate, 1-Boc-1,2,3,6-tetrahydropyridine, tert-Butyl 1,2,3,6-Tetrahydropyridine-1-carboxylate, 1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1,2,3,6-Tetrahydropyridine-1-carboxylic acid tert-Butyl ester

054030-1G 054030-5G
101905-896EA 21.54 USD
101905-896 101905-898
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester ≥97%
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
MDL Number: MFCD04972245
CAS Number: 85838-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR