5-Chloropyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

054028-1G 054028-5G
101905-890EA 46.15 USD
101905-890 101905-892
5-Chloropyrazine-2-carboxylic acid ≥95%
5-Chloropyrazine-2-carboxylic acid

Hazard: Irritant/Store in Freezer

Formula: C5H3ClN2O2
MDL Number: MFCD09033269
CAS Number: 36070-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR