2,6-Dibromo-3-pyridinecarboxylic acid ≥98%

Supplier: Matrix Scientific

054022-1G 054022-5G
101905-876EA 44.62 USD
101905-876 101905-878
2,6-Dibromo-3-pyridinecarboxylic acid ≥98%
2,6-Dibromo-3-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₆H₃Br₂NO₂
MDL Number: MFCD11846496
CAS Number: 55304-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR