2,6-Dibromo-3-iodopyridine ≥98%

Supplier: Matrix Scientific

054021-5G 054021-25G
101905-872EA 121.54 USD
101905-872 101905-874
2,6-Dibromo-3-iodopyridine ≥98%
2,6-Dibromo-3-iodopyridine

Hazard: Irritant

Formula: C₅H₂Br₂IN
MDL Number: MFCD13185520
CAS Number: 1032582-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR