Methyl-2-azabicyclo[2.2.1]heptane-3-carboxylate hydrochloride

Supplier: Matrix Scientific

054019-500MG 054019-2.5G
101905-868EA 461.54 USD
101905-868 101905-870
Methyl-2-azabicyclo[2.2.1]heptane-3-carboxylate hydrochloride
Methyl-2-azabicyclo[2.2.1]heptane-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₄ClNO₂
MDL Number: MFCD16631258
CAS Number: 88259-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR