2-Azabicyclo[2.2.1]hept-3-ylmethanol

Supplier: Matrix Scientific

054018-500MG 054018-2.5G
101905-864EA 384.62 USD
101905-864 101905-866
2-Azabicyclo[2.2.1]hept-3-ylmethanol
2-Azabicyclo[2.2.1]hept-3-ylmethanol

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD24390036
CAS Number: 347381-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR