2-Azabicyclo[2.2.1]heptane-3-carboxylic acid
Supplier: Matrix Scientific
054017-500MG
054017-2.5G
101905-860EA
364.62
USD
101905-860
101905-862
2-Azabicyclo[2.2.1]heptane-3-carboxylic acid
2-Azabicyclo[2.2.1]heptane-3-carboxylic acid
Hazard: Irritant
Formula:
C₇H₁₁NO₂ |
MDL Number:
MFCD07440107 CAS Number: 88260-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...