2-(4-Fluorophenyl)-2-methylpropanoic acid

Supplier: Matrix Scientific

054015-500MG 054015-2.5G
101905-852EA 187.69 USD
101905-852 101905-854
2-(4-Fluorophenyl)-2-methylpropanoic acid
2-(4-Fluorophenyl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD11036916
CAS Number: 93748-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR