2-(3-Fluorophenyl)-2-methylpropanoic acid

Supplier: Matrix Scientific

054014-500MG 054014-2.5G
101905-848EA 340 USD
101905-848 101905-850
2-(3-Fluorophenyl)-2-methylpropanoic acid
2-(3-Fluorophenyl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD11036911
CAS Number: 93748-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR