2-(4-Methoxyphenyl)-2-methylpropanoic acid

Supplier: Matrix Scientific

054013-500MG 054013-2.5G
101905-844EA 181.54 USD
101905-844 101905-846
2-(4-Methoxyphenyl)-2-methylpropanoic acid
2-(4-Methoxyphenyl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₃
MDL Number: MFCD11036946
CAS Number: 2955-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR