2-(3-Methoxyphenyl)-2-methylpropanoic acid
Supplier: Matrix Scientific
054012-500MG
054012-2.5G
101905-840EA
307.69
USD
101905-840
101905-842
2-(3-Methoxyphenyl)-2-methylpropanoic acid
2-(3-Methoxyphenyl)-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₄O₃ |
MDL Number:
MFCD09414710 CAS Number: 17653-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...