2-(2-Furyl)-2-methylpropanoic acid
Supplier: Matrix Scientific
054010-500MG
054010-2.5G
101905-832EA
364.62
USD
101905-832
101905-834
2-(2-Furyl)-2-methylpropanoic acid
2-(2-Furyl)-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₈H₁₀O₃ |
MDL Number:
MFCD13563036 CAS Number: 24954-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...