2-Methyl-2-[4-(trifluoromethyl)phenyl]propanoic acid

Supplier: Matrix Scientific

054006-500MG 054006-2.5G
101905-816EA 229.23 USD
101905-816 101905-818
2-Methyl-2-[4-(trifluoromethyl)phenyl]propanoic acid
2-Methyl-2-[4-(trifluoromethyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₁F₃O₂
MDL Number: MFCD10703524
CAS Number: 32445-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR