2-Methyl-2-[3-(trifluoromethyl)phenyl]propanoic acid

Supplier: Matrix Scientific

054005-500MG 054005-2.5G
101905-812EA 461.54 USD
101905-812 101905-814
2-Methyl-2-[3-(trifluoromethyl)phenyl]propanoic acid
2-Methyl-2-[3-(trifluoromethyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₁F₃O₂
MDL Number: MFCD11036961
CAS Number: 254895-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR