2-(1-Azepanyl)-5-pyrimidinecarboxylic acid

Supplier: Matrix Scientific

053995-500MG 053995-2.5G
101905-780EA 364.62 USD
101905-780 101905-782
2-(1-Azepanyl)-5-pyrimidinecarboxylic acid
2-(1-Azepanyl)-5-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD13563029
CAS Number: 1220027-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR