6-(1-Azepanyl)-2-pyrazinecarboxylic acid

Supplier: Matrix Scientific

053994-500MG 053994-2.5G
101905-776EA 364.62 USD
101905-776 101905-778
6-(1-Azepanyl)-2-pyrazinecarboxylic acid
6-(1-Azepanyl)-2-pyrazinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD01412252
CAS Number: 380638-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR