6-(4-Morpholinyl)-2-pyrazinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 6-(4-Morpholinyl)pyrazine-2-carboxylic acid

053993-500MG 053993-2.5G
101905-772EA 364.62 USD
101905-772 101905-774
6-(4-Morpholinyl)-2-pyrazinecarboxylic acid
6-(4-Morpholinyl)-2-pyrazinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₃
MW: 209.21 g/mol
MDL Number: MFCD00233274
CAS Number: 40262-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR