6-(Diethylamino)-2-pyrazinecarboxylic acid

Supplier: Matrix Scientific

053991-500MG 053991-2.5G
101905-764EA 364.62 USD
101905-764 101905-766
6-(Diethylamino)-2-pyrazinecarboxylic acid
6-(Diethylamino)-2-pyrazinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₃N₃O₂
MDL Number: MFCD05721823
CAS Number: 40262-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR