6-(Dimethylamino)-2-pyrazinecarboxylic acid

Supplier: Matrix Scientific

053990-500MG 053990-2.5G
101905-760EA 364.62 USD
101905-760 101905-762
6-(Dimethylamino)-2-pyrazinecarboxylic acid
6-(Dimethylamino)-2-pyrazinecarboxylic acid

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD05721827
CAS Number: 40262-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR