6-Methoxy-2-pyrazinecarboxylic acid

Supplier: Matrix Scientific

053989-500MG 053989-2.5G
101905-756EA 84.62 USD
101905-756 101905-758
6-Methoxy-2-pyrazinecarboxylic acid
6-Methoxy-2-pyrazinecarboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00233277
CAS Number: 24005-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR