4-[2-(3-Piperidinyl)ethyl]morpholine

Supplier: Matrix Scientific

053986-500MG 053986-2.5G
101905-744EA 364.62 USD
101905-744 101905-746
4-[2-(3-Piperidinyl)ethyl]morpholine
4-[2-(3-Piperidinyl)ethyl]morpholine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O
MDL Number: MFCD13563026
CAS Number: 1219979-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR