1-[2-(3-Piperidinyl)ethyl]piperidine

Supplier: Matrix Scientific

053985-500MG 053985-2.5G
101905-740EA 364.62 USD
101905-740 101905-742
1-[2-(3-Piperidinyl)ethyl]piperidine
1-[2-(3-Piperidinyl)ethyl]piperidine

Hazard: Irritant

Formula: C₁₂H₂₄N₂
MW: 196.34 g/mol
MDL Number: MFCD13563025
CAS Number: 122373-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR