2-[1-(4-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid
Supplier: Matrix Scientific
053984-500MG
053984-2.5G
101905-736EA
364.62
USD
101905-736
101905-738
2-[1-(4-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid
2-[1-(4-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD13563024 CAS Number: 1220027-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...