2-[1-(2-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid

Supplier: Matrix Scientific

053983-500MG 053983-2.5G
101905-732EA 364.62 USD
101905-732 101905-734
2-[1-(2-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid
2-[1-(2-Pyridinylmethyl)-3-pyrrolidinyl]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD13563023
CAS Number: 1220027-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR