2-(1-Benzyl-3-pyrrolidinyl)ethylamine

Supplier: Matrix Scientific

053982-500MG 053982-2.5G
101905-728EA 364.62 USD
101905-728 101905-730
2-(1-Benzyl-3-pyrrolidinyl)ethylamine
2-(1-Benzyl-3-pyrrolidinyl)ethylamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD03790943
CAS Number: 130927-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR