2-(1-Propyl-3-pyrrolidinyl)ethylamine

Supplier: Matrix Scientific

053980-500MG 053980-2.5G
101905-724EA 364.62 USD
101905-724 101905-726
2-(1-Propyl-3-pyrrolidinyl)ethylamine
2-(1-Propyl-3-pyrrolidinyl)ethylamine

Hazard: Irritant

Formula: C₉H₂₀N₂
MDL Number: MFCD13563022
CAS Number: 1219979-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR