2-[1-(Tetrahydro-2-furanylmethyl)-3-pyrrolidinyl]acetic acid

Supplier: Matrix Scientific

053977-500MG 053977-2.5G
101905-716EA 364.62 USD
101905-716 101905-718
2-[1-(Tetrahydro-2-furanylmethyl)-3-pyrrolidinyl]acetic acid
2-[1-(Tetrahydro-2-furanylmethyl)-3-pyrrolidinyl]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD13563021
CAS Number: 1220036-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR